Skip to Content
MilliporeSigma

AABH9BD8B8EA

AA BLOCKS, INC.

Nordoxepin

Synonym(s):

(3E)-3-dibenzo[b,e]oxepin-11(6H)-ylidene-N-methyl-1-propanamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C18H19O1N1
CAS Number:
Molecular Weight:
265.35
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C18H19NO/c1-19-12-6-10-16-15-8-3-2-7-14(15)13-20-18-11-5-4-9-17(16)18/h2-5,7-11,19H,6,12-13H2,1H3/b16-10+

Inchi Key

HVKCEFHNSNZIHO-MHWRWJLKSA-N

Storage temp.

-20C

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library