Skip to Content
MilliporeSigma

AABH9BD8EA22

AA BLOCKS, INC.

2,8,9-Trimethyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane hydrochloride

Synonym(s):

2,8,9-trimethyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H22N4Cl1P1
CAS Number:
Molecular Weight:
252.73
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H21N4P.ClH/c1-10-4-7-13-8-5-11(2)14(10)12(3)6-9-13;/h4-9H2,1-3H3;1H

Inchi Key

QKTVLAVMLZQIEI-UHFFFAOYSA-N

Purity

96%

Storage temp.

N/A

Country of Origin

US



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library