Skip to Content
MilliporeSigma

AABH9BD97E83

AA BLOCKS, INC.

L-Fucose-1-phosphate disodium salt

Synonym(s):

sodium (3S,4R,5S,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl phosphate

Select a Size

Change View

About This Item

Linear Formula:
C6H11O8P1Na2
CAS Number:
Molecular Weight:
288.1
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H13O8P.2Na/c1-2-3(7)4(8)5(9)6(13-2)14-15(10,11)12;;/h2-9H,1H3,(H2,10,11,12);;/q;2*+1/p-2/t2-,3+,4+,5-,6?;;/m0../s1

Inchi Key

JPJQYRZIJVIUSX-MOQCCXJMSA-L

Purity

95%

Country of Origin

US

Storage temp.

N/A





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library