Skip to Content
MilliporeSigma

ATEH9BCAAFA2

AstaTech, Inc.

ACETOXIME BENZOATE

Synonym(s):

ethenylamino benzoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H9NO2/c1-2-10-12-9(11)8-6-4-3-5-7-8/h2-7,10H,1H2

Inchi Key

DJJQTOBCGKKISP-UHFFFAOYSA-N

Purity

95%

Physical Form

White to Yellow Solid

Shipping Temperature

Cold box

Storage temp.

-20 C

Country of Origin

CN



Signal Word

Warning



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library