Skip to Content
MilliporeSigma

BDPH9BCC25A8

Bide Pharmatech

1-(2-Bromo-4-(trifluoromethyl)phenyl)ethanone

Synonym(s):

1-[2-bromo-4-(trifluoromethyl)phenyl]ethanone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H6O1Br1F3
CAS Number:
Molecular Weight:
267.05
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H6BrF3O/c1-5(14)7-3-2-6(4-8(7)10)9(11,12)13/h2-4H,1H3

Inchi Key

HBGXHUPYBNIJTQ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Colorless to Yellow Liquid



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library