Skip to Content
MilliporeSigma

BDPH9BCC2E8B

Bide Pharmatech

4,4,5,5-Tetramethyl-2-(2,4,5-trimethylthiophen-3-yl)-1,3,2-dioxaborolane

Synonym(s):

4,4,5,5-tetramethyl-2-(2,4,5-trimethylthiophen-3-yl)-1,3,2-dioxaborolane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C13H21O2S1B1
CAS Number:
Molecular Weight:
252.19
MDL number:


Inchi Code

1S/C13H21BO2S/c1-8-9(2)17-10(3)11(8)14-15-12(4,5)13(6,7)16-14/h1-7H3

Inchi Key

HDZMOJWLMHUYBK-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid or semi-solid



Still not finding the right product?


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library