Skip to Content
MilliporeSigma

BDPH9BCC3012

Bide Pharmatech

6-(Dimethylamino)nicotinonitrile

Synonym(s):

6-(dimethylamino)nicotinonitrile

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H9N3
CAS Number:
Molecular Weight:
147.18
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H9N3/c1-11(2)8-4-3-7(5-9)6-10-8/h3-4,6H,1-2H3

Inchi Key

YIZXYFJKSCABES-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 6-(Dimethylamino)nicotinonitrile



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library