Skip to Content
MilliporeSigma

BDPH9BCC360B

Bide Pharmatech

3-Chloro-5-methylpyrazine-2-carboxamide

Synonym(s):

3-chloro-5-methylpyrazine-2-carboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H6O1N3Cl1
CAS Number:
Molecular Weight:
171.59
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H6ClN3O/c1-3-2-9-4(6(8)11)5(7)10-3/h2H,1H3,(H2,8,11)

Inchi Key

JEDOBXCGHBZIDZ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library