Skip to Content
MilliporeSigma

BDPH9BCC3CF5

Bide Pharmatech

Methyl 5-bromo-2-ethylbenzoate

Synonym(s):

methyl 5-bromo-2-ethylbenzoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H11O2Br1
CAS Number:
Molecular Weight:
243.1
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H11BrO2/c1-3-7-4-5-8(11)6-9(7)10(12)13-2/h4-6H,3H2,1-2H3

Inchi Key

NYMZTCOVKDJUKT-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Colorless to Yellow Liquid



Still not finding the right product?

Explore all of our products under Methyl 5-bromo-2-ethylbenzoate


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library