Skip to Content
MilliporeSigma

BDPH9BCC3E4E

Bide Pharmatech

N,N'''-1,6-Hexanediylbis(N'-cyanoguanidine)

Synonym(s):

N-cyano-N'-[6-(N'-cyanocarbamimidamido)hexyl]guanidine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H18N8
CAS Number:
Molecular Weight:
250.31
MDL number:


Inchi Code

1S/C10H18N8/c11-7-17-9(13)15-5-3-1-2-4-6-16-10(14)18-8-12/h1-6H2,(H3,13,15,17)(H3,14,16,18)

Inchi Key

YXZZOMVBHPCKMM-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

White to off-white powder or crystals



Still not finding the right product?


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library