Skip to Content
MilliporeSigma

BDPH9BCC42D3

Bide Pharmatech

[4-(Ethanesulfonyl)phenyl]methanamine hydrochloride

Synonym(s):

(4-(ethylsulfonyl)phenyl)methanamine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H14O2N1Cl1S1
CAS Number:
Molecular Weight:
235.73
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H13NO2S.ClH/c1-2-13(11,12)9-5-3-8(7-10)4-6-9;/h3-6H,2,7,10H2,1H3;1H

Inchi Key

KXPRDHZTUZNMQT-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library