Skip to Content
MilliporeSigma

BDPH9BCC47DF

Bide Pharmatech

2-(Phenylamino)nicotinic acid

Synonym(s):

2-anilinonicotinic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H10O2N2
CAS Number:
Molecular Weight:
214.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H10N2O2/c15-12(16)10-7-4-8-13-11(10)14-9-5-2-1-3-6-9/h1-8H,(H,13,14)(H,15,16)

Inchi Key

UUMMTMQODCACRH-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 2-(Phenylamino)nicotinic acid


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library