Skip to Content
MilliporeSigma

BDPH9BCC4C31

Bide Pharmatech

2-Methyl-4-(trifluoromethoxy)phenol

Synonym(s):

2-methyl-4-(trifluoromethoxy)phenol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H7O2F3
CAS Number:
Molecular Weight:
192.14
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H7F3O2/c1-5-4-6(2-3-7(5)12)13-8(9,10)11/h2-4,12H,1H3

Inchi Key

QMIXWHAIFDQEDH-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Country of Origin

CN

Physical Form

Solid or semi-solid or liquid

Storage temp.

Sealed in dry, 2-8°C



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library