Skip to Content
MilliporeSigma

BDPH9BCC6029

Bide Pharmatech

3-Bromo-5,6,7,8-tetrahydroimidazo[1,5-a]pyrazine hydrochloride

Synonym(s):

3-bromo-5,6,7,8-tetrahydroimidazo[1,5-a]pyrazine hydrochloride

Select a Size

Change View

About This Item

Linear Formula:
C6H9N3Cl1Br1
CAS Number:
Molecular Weight:
238.51
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H8BrN3.ClH/c7-6-9-4-5-3-8-1-2-10(5)6;/h4,8H,1-3H2;1H

Inchi Key

FNKYKFZYHXJZRI-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library