Skip to Content

BDPH9BCC66CB

Bide Pharmatech

1-Ethyl-2-oxo-1,2-dihydropyridine-4-carboxylic acid

Synonym(s):

1-ethyl-2-oxo-1,2-dihydro-4-pyridinecarboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H9O3N1
CAS Number:
Molecular Weight:
167.16
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H9NO3/c1-2-9-4-3-6(8(11)12)5-7(9)10/h3-5H,2H2,1H3,(H,11,12)

Inchi Key

MZLMUTMYNUZZTC-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library