Inchi Code
1S/C12H23NO4/c1-12(2,3)17-11(16)13(4)9-7-5-6-8-10(14)15/h5-9H2,1-4H3,(H,14,15)
Inchi Key
NWRXFDQJTBAVMC-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Sealed in dry, room temperature
Country of Origin
CN
Physical Form
Solid or viscous liquid or liquid
Still not finding the right product?
Explore all of our products under 6-((tert-Butoxycarbonyl)(methyl)amino)hexanoic acid
hstatements
signalword
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
