Skip to Content
MilliporeSigma

BDPH9BCC9CBE

Bide Pharmatech

2-(1H-Imidazol-2-yl)propan-2-ol

Synonym(s):

2-(1H-imidazol-2-yl)-2-propanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H10O1N2
CAS Number:
Molecular Weight:
126.16
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H10N2O/c1-6(2,9)5-7-3-4-8-5/h3-4,9H,1-2H3,(H,7,8)

Inchi Key

CJYJVIFSBZSJBN-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 2-(1H-Imidazol-2-yl)propan-2-ol


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library