Skip to Content
MilliporeSigma

BDPH9BCCBC46

Bide Pharmatech

3-(4-Bromophenyl)morpholine hydrochloride

Synonym(s):

3-(4-bromophenyl)morpholine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H13O1N1Cl1Br1
CAS Number:
Molecular Weight:
278.58
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H12BrNO.ClH/c11-9-3-1-8(2-4-9)10-7-13-6-5-12-10;/h1-4,10,12H,5-7H2;1H

Inchi Key

GWQXSBSWSBIWEV-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library