Skip to Content
MilliporeSigma

BDPH9BCCC16D

Bide Pharmatech

3-(Aminomethyl)oxetan-3-amine dihydrochloride

Synonym(s):

3-(aminomethyl)oxetan-3-amine dihydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C4H12O1N2Cl2
CAS Number:
Molecular Weight:
175.06
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C4H10N2O.2ClH/c5-1-4(6)2-7-3-4;;/h1-3,5-6H2;2*1H

Inchi Key

UMVLEUYAOATFGB-UHFFFAOYSA-N

Shipping Temperature

Cold-chain transportation

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library