Skip to Content
MilliporeSigma

BDPH9BCCCB59

Bide Pharmatech

(2-(Benzyloxy)phenyl)methanol

Synonym(s):

[2-(benzyloxy)phenyl]methanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C14H14O2
CAS Number:
Molecular Weight:
214.26
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C14H14O2/c15-10-13-8-4-5-9-14(13)16-11-12-6-2-1-3-7-12/h1-9,15H,10-11H2

Inchi Key

XRZMRABYCSOXIZ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid or Semi-solid or liquid or lump



Still not finding the right product?

Explore all of our products under (2-(Benzyloxy)phenyl)methanol


hstatements

Precautionary Statements

Signal Word

Danger

Pictograms

Corrosion


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library