Skip to Content
MilliporeSigma

BDPH9BCCDA99

Bide Pharmatech

2-(2,6-Dimethylpiperidin-1-yl)ethanol

Synonym(s):

2-(2,6-dimethyl-1-piperidinyl)ethanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H19O1N1
CAS Number:
Molecular Weight:
157.26
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H19NO/c1-8-4-3-5-9(2)10(8)6-7-11/h8-9,11H,3-7H2,1-2H3

Inchi Key

NJBXJPLWOZJAJJ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library