Skip to Content
MilliporeSigma

BDPH9BCCDB88

Bide Pharmatech

3-(3-(Trifluoromethyl)-1H-pyrazol-1-yl)butanoic acid

Synonym(s):

3-[3-(trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H9O2N2F3
CAS Number:
Molecular Weight:
222.17
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H9F3N2O2/c1-5(4-7(14)15)13-3-2-6(12-13)8(9,10)11/h2-3,5H,4H2,1H3,(H,14,15)

Inchi Key

OWRBXDUQRMCGBL-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid or semi-solid or liquid



Still not finding the right product?


hstatements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library