Skip to Content
MilliporeSigma

BDPH9BCCE1B5

Bide Pharmatech

5-(Chloromethyl)furan-2-carbaldehyde

Synonym(s):

5-(chloromethyl)-2-furaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H5O2Cl1
CAS Number:
Molecular Weight:
144.56
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H5ClO2/c7-3-5-1-2-6(4-8)9-5/h1-2,4H,3H2

Inchi Key

KAZRCBVXUOCTIO-UHFFFAOYSA-N

Shipping Temperature

Cold-chain transportation

Purity

95%

Storage temp.

Inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

Solid or semi-solid or lump or liquid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library