Skip to Content
MilliporeSigma

BDPH9BCCF54D

Bide Pharmatech

1-(2-(Difluoromethoxy)phenyl)ethan-1-amine

Synonym(s):

1-[2-(difluoromethoxy)phenyl]ethylamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H11O1N1F2
CAS Number:
Molecular Weight:
187.19
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H11F2NO/c1-6(12)7-4-2-3-5-8(7)13-9(10)11/h2-6,9H,12H2,1H3

Inchi Key

XZXJJKFVKBSRME-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library