Skip to Content
MilliporeSigma

BDPH9BCCFACE

Bide Pharmatech

(4-(Cyclohexylmethoxy)phenyl)boronic acid

Synonym(s):

4-(cyclohexylmethoxy)phenylboronic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C13H19O3B1
CAS Number:
Molecular Weight:
234.1
MDL number:


Inchi Code

1S/C13H19BO3/c15-14(16)12-6-8-13(9-7-12)17-10-11-4-2-1-3-5-11/h6-9,11,15-16H,1-5,10H2

Inchi Key

KXOJKGOXRZMOBU-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library