Skip to Content
MilliporeSigma

BDPH9BCD06B1

Bide Pharmatech

4-(4-Bromo-1H-pyrazol-3-yl)pyridine

Synonym(s):

4-(4-bromo-1H-pyrazol-3-yl)pyridine

Select a Size

Change View

About This Item

Linear Formula:
C8H6N3Br1
CAS Number:
Molecular Weight:
224.06
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H6BrN3/c9-7-5-11-12-8(7)6-1-3-10-4-2-6/h1-5H,(H,11,12)

Inchi Key

LKKNQJJSEMQTHQ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

White to Yellow Solid



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library