Skip to Content
MilliporeSigma

BDPH9BCD0714

Bide Pharmatech

5-(4-Isopropylphenyl)-1,3,4-thiadiazol-2-amine

Synonym(s):

5-(4-isopropylphenyl)-1,3,4-thiadiazol-2-amine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H13N3S1
CAS Number:
Molecular Weight:
219.31
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H13N3S/c1-7(2)8-3-5-9(6-4-8)10-13-14-11(12)15-10/h3-7H,1-2H3,(H2,12,14)

Inchi Key

QEKWMJDRQOKJJS-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Country of Origin

CN

Physical Form

White to light-yellow powder or crystals



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library