Skip to Content
MilliporeSigma

BDPH9BCD09F5

Bide Pharmatech

2-Amino-N-(5-chloropyridin-2-yl)-5-methoxybenzamide

Synonym(s):

2-amino-N-(5-chloro-2-pyridinyl)-5-methoxybenzamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C13H12O2N3Cl1
CAS Number:
Molecular Weight:
277.71
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C13H12ClN3O2/c1-19-9-3-4-11(15)10(6-9)13(18)17-12-5-2-8(14)7-16-12/h2-7H,15H2,1H3,(H,16,17,18)

Inchi Key

GCCIHZVIPXGAPR-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library