Skip to Content

BDPH9BCD0A84

Bide Pharmatech

Methyl 2-(4-(2-bromoethyl)phenyl)acetate

Synonym(s):

methyl 2-(4-(2-bromoethyl)phenyl)acetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H13O2Br1
Molecular Weight:
257.13
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H13BrO2/c1-14-11(13)8-10-4-2-9(3-5-10)6-7-12/h2-5H,6-8H2,1H3

Inchi Key

QXRJAWGFXUBQGW-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library