Skip to Content
MilliporeSigma

BDPH9BCD0B9A

Bide Pharmatech

S-(3-Oxopropyl) ethanethioate

Synonym(s):

S-(3-oxopropyl) ethanethioate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H8O2S1
CAS Number:
Molecular Weight:
132.18
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H8O2S/c1-5(7)8-4-2-3-6/h3H,2,4H2,1H3

Inchi Key

LLGHPZSANAZMFY-UHFFFAOYSA-N

Shipping Temperature

Cold-chain transportation

Purity

95%

Country of Origin

CN

Physical Form

Liquid

Storage temp.

Inert atmosphere, store in freezer, under -20°C



Still not finding the right product?

Explore all of our products under S-(3-Oxopropyl) ethanethioate



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library