Skip to Content
MilliporeSigma

BDPH9BCD0DEA

Bide Pharmatech

1-(3-Bromophenyl)-2-hydroxyethan-1-one

Synonym(s):

1-(3-bromophenyl)-2-hydroxyethanone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H7O2Br1
CAS Number:
Molecular Weight:
215.05
MDL number:


Inchi Code

1S/C8H7BrO2/c9-7-3-1-2-6(4-7)8(11)5-10/h1-4,10H,5H2

Inchi Key

RSWPGWSYMLTIOH-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

96%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library