Skip to Content
MilliporeSigma

BDPH9BCD14DA

Bide Pharmatech

(11bR)-N-(2,6-Bis(4-methoxyphenyl)-4-oxido-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide

Synonym(s):

N-(2,6-bis(4-methoxyphenyl)-4-oxido-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C35H33O7N1S1P1F3
CAS Number:
Molecular Weight:
699.68
MDL number:


Inchi Code

1S/C35H33F3NO7PS/c1-43-25-15-11-21(12-16-25)29-19-23-7-3-5-9-27(23)31-32-28-10-6-4-8-24(28)20-30(22-13-17-26(44-2)18-14-22)34(32)46-47(40,45-33(29)31)39-48(41,42)35(36,37)38/h11-20H,3-10H2,1-2H3,(H,39,40)

Inchi Key

RXLOACMJQMXVPU-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid






Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library