Skip to Content

BDPH9BCD388F

Bide Pharmatech

2-(Naphthalen-1-yl)acetonitrile

Synonym(s):

1-naphthylacetonitrile

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H9N1
CAS Number:
Molecular Weight:
167.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H9N/c13-9-8-11-6-3-5-10-4-1-2-7-12(10)11/h1-7H,8H2

Inchi Key

OQRMWUNUKVUHQO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Liquid or Solid or Semi-solid



Still not finding the right product?

Explore all of our products under 2-(Naphthalen-1-yl)acetonitrile


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library