Inchi Code
1S/C17H17NO3/c1-12(19)18-16(17(20)21)11-13-7-9-15(10-8-13)14-5-3-2-4-6-14/h2-10,16H,11H2,1H3,(H,18,19)(H,20,21)
Inchi Key
HDNGVBPPMAZUMI-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Sealed in dry, room temperature
Country of Origin
CN
Physical Form
Powder or crystals
Still not finding the right product?
Explore all of our products under 3-([1,1'-Biphenyl]-4-yl)-2-acetamidopropanoic acid
hstatements
signalword
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
