Skip to Content
MilliporeSigma

BDPH9BCD40C6

Bide Pharmatech

2,3-DIaminobenzoic acid hydrochloride

Synonym(s):

2,3-diaminobenzoic acid hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H9O2N2Cl1
Molecular Weight:
188.61
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H8N2O2.ClH/c8-5-3-1-2-4(6(5)9)7(10)11;/h1-3H,8-9H2,(H,10,11);1H

Inchi Key

OMWNWENDHSMOPJ-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library