Skip to Content

BDPH9BCD448C

Bide Pharmatech

4-Aminobutan-2-one hydrochloride

Synonym(s):

4-amino-2-butanone hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C4H10O1N1Cl1
Molecular Weight:
123.58
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C4H9NO.ClH/c1-4(6)2-3-5;/h2-3,5H2,1H3;1H

Inchi Key

CRYWPNCOLVABFJ-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library