Skip to Content
MilliporeSigma

BDPH9BCD4A44

Bide Pharmatech

1-(2,2-Dimethoxyethyl)-1H-pyrazole

Synonym(s):

1-(2,2-dimethoxyethyl)-1H-pyrazole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H12O2N2
CAS Number:
Molecular Weight:
156.18
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H12N2O2/c1-10-7(11-2)6-9-5-3-4-8-9/h3-5,7H,6H2,1-2H3

Inchi Key

YIEUOHXVZSLNAS-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under 1-(2,2-Dimethoxyethyl)-1H-pyrazole



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library