Skip to Content
MilliporeSigma

BDPH9BCD4AAD

Bide Pharmatech

Hexadecyltrimethoxysilane

Synonym(s):

hexadecyl(trimethoxy)silane

Select a Size

Change View

About This Item

Linear Formula:
C19H42O3Si1
CAS Number:
Molecular Weight:
346.63
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C19H42O3Si/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-23(20-2,21-3)22-4/h5-19H2,1-4H3

Inchi Key

RSKGMYDENCAJEN-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Form: liquid / Colour: Colorless - Almost colorless



Still not finding the right product?

Explore all of our products under Hexadecyltrimethoxysilane


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library