Skip to Content

BDPH9BCD4AB2

Bide Pharmatech

1-Ethyl-1H-pyrazole-5-carbaldehyde

Synonym(s):

1-ethyl-1H-pyrazole-5-carbaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H8O1N2
CAS Number:
Molecular Weight:
124.14
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H8N2O/c1-2-8-6(5-9)3-4-7-8/h3-5H,2H2,1H3

Inchi Key

SKUJVOCRQZNYEX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Colorless to Yellow Liquid



Still not finding the right product?

Explore all of our products under 1-Ethyl-1H-pyrazole-5-carbaldehyde


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library