Skip to Content

BDPH9BCD58CF

Bide Pharmatech

4-Bromo-2-(carboxymethyl)benzoic acid

Synonym(s):

4-bromo-2-(carboxymethyl)benzoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H7O4Br1
CAS Number:
Molecular Weight:
259.06
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H7BrO4/c10-6-1-2-7(9(13)14)5(3-6)4-8(11)12/h1-3H,4H2,(H,11,12)(H,13,14)

Inchi Key

WDKVNGVXRHWUEQ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library