Skip to Content

BDPH9BCD7C04

Bide Pharmatech

5-Bromo-6-hydroxy-2-methylpyrimidin-4(1H)-one

Synonym(s):

5-bromo-6-hydroxy-2-methyl-4(1H)-pyrimidinone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H5O2N2Br1
CAS Number:
Molecular Weight:
205.01
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Storage temp.

Sealed in dry, 2-8°C

Inchi Code

1S/C5H5BrN2O2/c1-2-7-4(9)3(6)5(10)8-2/h3H,1H2,(H,7,9)(H,8,10)

Inchi Key

ZAKXWSORBVRPGY-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library