Skip to Content
MilliporeSigma

BDPH9BCD8353

Bide Pharmatech

4-Bromo-2-chloro-1-(trifluoromethoxy)benzene

Synonym(s):

4-bromo-2-chloro-1-(trifluoromethoxy)benzene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H3O1Cl1Br1F3
CAS Number:
Molecular Weight:
275.45
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H3BrClF3O/c8-4-1-2-6(5(9)3-4)13-7(10,11)12/h1-3H

Inchi Key

WSENYRJAJFHXQR-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Light-yellow to yellow liquid



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library