Skip to Content

BDPH9BCD83A2

Bide Pharmatech

3-Fluorobicyclo[1.1.1]pentane-1-carboxylic acid

Synonym(s):

3-fluorobicyclo[1.1.1]pentane-1-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H7O2F1
CAS Number:
Molecular Weight:
130.12
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H7FO2/c7-6-1-5(2-6,3-6)4(8)9/h1-3H2,(H,8,9)

Inchi Key

CQOUAQUBJPKQON-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Country of Origin

CN

Physical Form

White to Yellow Solid

Storage temp.

Sealed in dry, 2-8°C



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library