Skip to Content
MilliporeSigma

BDPH9BCD8B04

Bide Pharmatech

1-(4-Bromophenyl)-4-chlorobutan-1-one

Synonym(s):

1-(4-bromophenyl)-4-chloro-1-butanone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H10O1Cl1Br1
CAS Number:
Molecular Weight:
261.55
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H10BrClO/c11-9-5-3-8(4-6-9)10(13)2-1-7-12/h3-6H,1-2,7H2

Inchi Key

WJKPUMBLABGUCQ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid or semi-solid or lump or liquid



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library