Skip to Content
MilliporeSigma

BDPH9BCD8C7F

Bide Pharmatech

3-Bromo-1-(piperidin-1-yl)propan-1-one

Synonym(s):

1-(3-bromopropanoyl)piperidine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H14O1N1Br1
CAS Number:
Molecular Weight:
220.11
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H14BrNO/c9-5-4-8(11)10-6-2-1-3-7-10/h1-7H2

Inchi Key

QLCFKORONJMTNL-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library