Skip to Content
MilliporeSigma

BDPH9BCD8FF8

Bide Pharmatech

7-((2-(2,6-Dioxopiperidin-3-yl)-1-oxoisoindolin-4-yl)amino)heptanoic acid

Synonym(s):

7-((2-(6-hydroxy-2-oxo-2,3,4,5-tetrahydropyridin-3-yl)-1-oxoisoindolin-4-yl)amino)heptanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C20H25O5N3
CAS Number:
Molecular Weight:
387.44
MDL number:


Inchi Code

1S/C20H25N3O5/c24-17-10-9-16(19(27)22-17)23-12-14-13(20(23)28)6-5-7-15(14)21-11-4-2-1-3-8-18(25)26/h5-7,16,21H,1-4,8-12H2,(H,25,26)(H,22,24,27)

Inchi Key

YEOPNJABOAOYEU-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library