Skip to Content
MilliporeSigma

BDPH9BCD96AD

Bide Pharmatech

2-(Vinyloxy)tetrahydro-2H-pyran

Synonym(s):

tetrahydro-2H-pyran-2-yl vinyl ether

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H12O2
CAS Number:
Molecular Weight:
128.17
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H12O2/c1-2-8-7-5-3-4-6-9-7/h2,7H,1,3-6H2

Inchi Key

BYUNYALHUMSCSA-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under 2-(Vinyloxy)tetrahydro-2H-pyran


hstatements

Signal Word

Danger

Pictograms

Flame


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library