Skip to Content
MilliporeSigma

BDPH9BCD9A93

Bide Pharmatech

5,7-Dichloropyrazolo[1,5-a]pyrimidine

Synonym(s):

5,7-dichloropyrazolo[1,5-a]pyrimidine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H3N3Cl2
CAS Number:
Molecular Weight:
188.02
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H3Cl2N3/c7-4-3-5(8)11-6(10-4)1-2-9-11/h1-3H

Inchi Key

JMTFWCYVZOFHLR-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

White to Pale-yellow to Yellow-brown Solid



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library