Skip to Content

BDPH9BCDA209

Bide Pharmatech

6-Fluoro-9H-pyrido[3,4-b]indole

Synonym(s):

6-fluoro-9H-beta-carboline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H7N2F1
CAS Number:
Molecular Weight:
186.19
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H7FN2/c12-7-1-2-10-9(5-7)8-3-4-13-6-11(8)14-10/h1-6,14H

Inchi Key

AHWZJZOSPUCFPC-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 6-Fluoro-9H-pyrido[3,4-b]indole



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library