Skip to Content
MilliporeSigma

BDPH9BCDA30D

Bide Pharmatech

1,2,3,4-Tetrahydro-1,6-naphthyridine

Synonym(s):

1,2,3,4-tetrahydro[1,6]naphthyridine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H10N2
CAS Number:
Molecular Weight:
134.18
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H10N2/c1-2-7-6-9-5-3-8(7)10-4-1/h3,5-6,10H,1-2,4H2

Inchi Key

ARCSZHIPWBDSGJ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library